Learn More
Electron Microscopy Sciences SEM Pin Stub Holder for 12 Zeiss 12.7 mm
Supplier: Electron Microscopy Sciences 7595212ZE
This 62 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to twelve standard 12.7 mm (1/2") pin stubs
Type: Zeiss with Built-in Dovetail