Learn More
Electron Microscopy Sciences SEM Mount Hitachi M4 1" Surface 9-Division, 1/4" height, Aluminum
Supplier: Electron Microscopy Sciences 75621D
Hitachi cylinder mount has a M4 tapered hole with female thread. Made from lathe finished ultra-pure aluminum. M4 tapered hole.
Head: 1"
Height: 6 mm