Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 9-Division 25 mm Head with 9 mm Pin, Aluminum, 3.2 mm Pin diameter
Supplier: Electron Microscopy Sciences 7518350
9 divisions surface min stub mounts are for use with FEI/Philips, ZEISS/LEO, Cambridge, Leica, Amray, Tescan and Camscan SEMs. Mounts are made from lathe finished ultra-pure aluminum.
Head: 1"
Pin height: 9 mm
Pin diameter: 3.2 mm