Learn More
Electron Microscopy Sciences SEM Cylinder Mount 45° Angle 5/8" Surface, Aluminum
Supplier: Electron Microscopy Sciences 7524110
Scanning electron microscope specimen mount with 45 degree angle. Made from lathe finished ultra-pure aluminum.
Head: 5/8"
Height: 3/8"