Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sellstrom™ Welding Magnifier Plates
$13.01
Specifications
Product Type | Magnifier Plate |
---|---|
For Use With (Application) | Welding |
Length (English) | 2 in. |
Material | Plastic |
Width (English) | 4.25 in. |
Catalog Number | Mfr. No. | Magnification Power | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Magnification Power | Price | Quantity & Availability | |||||
17-985-6D
|
Sellstrom™
18250 |
+2.50 |
Each for $13.01
|
|
17-985-6C
|
Sellstrom™
18200 |
+2.00 |
Each for $13.01
|
|
17-985-6A
|
Sellstrom™
18100 |
+1.00 |
Each for $13.01
|
|
17-985-6B
|
Sellstrom™
18150 |
+1.50 |
Each for $13.01
|
|
Specifications
Magnifier Plate | |
2 in. | |
4.25 in. |
Welding | |
Plastic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title