Learn More
Sellstrom™ Spill Control Station
Equipped for protection clean-up personnel
Supplier: Sellstrom™ 68000
Includes:
- 2 pairs of chemical splash goggles
- 15 sorbent pads
- 2 pairs of gloves (size Large)
- 15 hazardous waste bags and ties
- 2 total body Saranex Tyvek coveralls to provide head-to-toe protection
- Universal spill control pillows absorb and contain all types of acid, caustic, solvent or oil
Description
- Each station has 12 standard spill control pillows and 2 hydrofluoric acid pillows
- 11.5D × 15W × 30.75 in. (29.2 × 38.1 × 78.1cm)
Specifications
10-10 Spill Control Station | |
15 in. | |
30.75 in. | |
11.5 in. |
2 pairs of chemical splash goggles, 15 sorbent pads, 2 pairs of gloves (size large), 15 hazardous waste bags and ties, 2 total body Saranex Tyvek coveralls to provide head-to-toe protection | |
38 cm | |
78 cm | |
29.21 cm |