Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sellstrom™ Polycarbonate Filter Plates
Supplier: Sellstrom™ 16700 9
Specifications
Shade 9 | |
Sellstrom 290 Series Welding Hood | |
11.25 cm | |
5.25 in. |
Welding Filter Plate | |
4.5 in. | |
Polycarbonate | |
13.25 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title