Learn More
Strem, An Ascensus Company CAS# 7782-49-2. 50g. Selenium shot (99.99%). MFCD00134090
Supplier: Strem, An Ascensus Company 93341750G
CAS# 7782-49-2. 50g. Selenium shot (99.99%). MFCD00134090. Molecular Weight: 78.96. Molecular Formula: Se. Color/form: gray pellets (~2mm dia.) Strem# 93-3417. http://www.strem.com/catalog/v/93-3417/