Learn More
Norell Inc 5mm, 800MHz 7" SELECT SERIES NMR TUBE, 5 Tubes per pack
Supplier: Norell Inc S58007
Select Series NMR Tubes are ideal for cold refrigeration storage, work with cryo-probes and, variable temperature studies. Key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or under any other application where large temperature variations are required in the experiment.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.