Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alcor™ Seditrol™ Quality Control
Seditrol™ is a bi-level quality control designed to be used in the same manner as the patient sample.
Supplier: Alcor™ DSC01
Description
- A bi-level quality control, manufactured from human blood used to monitor the performance of erythrocyte sedimentation rate (ESR) procedures
- Used to monitor the performance of the laboratory's analyzer and the quality of the technique, as well as the effect environmental factors such as temperature, bench top vibration, and sample suitability may have on the quality of the test results
- No refrigeration required (store at room temperature between 18 to 30°C)
- 18 month shelf life from date of manufacture
- 31 day open vial stability or 40 QC runs in each tube
- Lot sequestering available upon request
Specifications
Strore at room temperature between 18 to 30°C | |
Human Blood | |
1 x 4.5 mL Level 1, 1 x 4.5 mL Level 2 | |
31 day open vial stability or 40 QC runs in each tube |
miniiSED™ automted ESR analyzer | |
Bi-level | |
Biological Source |
Safety and Handling
ShelfLife : 18 months from date of manufacture
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction