Learn More
AVANTOR MASTERFLEX (VWR LLC) 155759-43 Double open end polypropylene sediment cartridge; 50 m, 20 inch length
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0151242
Ideal for removing a broad range of deformable and nondeformable particles. Rigid design is ideal for filtering high-solids, higher-viscosity fluids that require higher differential pressures. Cartridge imparts no taste, odor, or color. Maximum temperature is 175¿F (79¿C).
Note: For optimal performance, change cartridges at 25 psid.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.