Learn More
Andwin Scientific Securline Lab Marker, 10/pk
Supplier: Andwin Scientific P122010PK
Securline Lab MarkersThis special-purpose writing instrument is designed to write on damp surfaces as easily as it writes on dry surfaces.Can be used on dry, wet or cold surfaces, and plastic or metal surfaces, such as petri dishes. Black quick-dry ink is designed not to bleed when subjected to water or alcohol during normal use. Can be washed off reusable surfaces. allowing surfaces to be marked repeatedly.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.