Learn More
IMEB, Inc Microscope Slide and Cassette Processing Pens, Black, 10 per Pack
Supplier: IMEB, Inc P1220
For cassettes and microscope slides. IMEB processing pens are resistant to alcohol, xylene, and cryogenic temperatures. Sturdy fine point nib makes thin easy to read lines. 10 per pack.
This item is not returnable.
View return policy