Learn More
3M™ Secure Click™ Organic Vapor/Acid Gas Respirator Cartridge
The Secure Click™ Organic Vapor/Acid Gas Cartridge is NIOSH-approved to help provide respiratory protection against certain organic vapors, chlorine, hydrogen chloride/sulfur dioxide, chlorine dioxide or hydrogen sulfide.
Supplier: 3M™ D8003
Description
- Secure Click™ cartridges utilize dual flow technology for easy breathing
- Secure Click™ connection style enables a quick, simple connection of cartridges
- Swept-back design for enhanced field of view and comfort
- For workplace/occupational use only
- Applications: Protection against certain organic vapors, chlorine, hydrogen chloride/sulfur dioxide, or chlorine dioxide or hydrogen sulfide
Specifications
Vapor | |
Bayonet | |
Secure Click | |
No | |
D8000 |
NIOSH Approved | |
Black | |
Cartridge | |
Secure Click Half Facepiece HF-800 Reusable Respirators | |
60 Cartridges/Pk. |