Learn More
Seca Corp 354 2-in-1 mobile baby scale and flat scale for toddlers
Supplier: Seca Corp 3541317004
Two scales in one! The seca 354 baby scale has a comfortable weighing tray and a fine graduation of 0.5 oz. In no time at all, it can be converted into a floor scale for children with a capacity of 44 lbs. The weighing tray and the base which serves as the floor scale are securely locked together, thus ensuring that the baby can be weighed safely and securely. A further advantage of the scale is its Breast-Milk-Intake-Function which determines how much milk the baby has consumed. The robust but lightweight scale is easy to transport and the user friendly buttons make it easy to operate. • Weigh babies lying down and toddlers standing up • Easily detach the weighing tray from the base to use as a flat scale up to 44 lbs capacity • Compact, robust and lightweight: ideal for mobile use • High-quality baby scale with calibration approval at an economical price • Wide range of functions: TARE, HOLD and Breast-Milk-Intake-Function (BMIF)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.