Learn More
Seca Corp 284 EMR-validated measuring station for height and weight
Supplier: Seca Corp 2841300109
seca 284 allows measuring and weighing to be done in on simple step. Other benefits include the high-capacity scale integrated in a stable glass platform. Measuring station is equipped with two displays: one for height from the headpiece and a touch-display that shows height, weight and BMI. • Measuring and weighing in one step • Precise 0.1 pound graduation • Backlit display in the headpiece shows the height • EMR-validated: transmit measured data directly to any electronic medical record system or printer • Capacity 660 lbs / 300 kg • Graduation 0.1 lb / 50 g • Functions: Pre-TARE, mother-child function, HOLD, Auto-HOLD, Auto-BMI, Auto-CLEAR, CLEAR, lbs/kg switch-over, inch/cm switch-over, damping • Accessories: Interface module seca 452, Panda seca 459 • Height measurement: Measuring range: 11 – 7 ft 2 / 30 – 220 cm • Graduation 1/8 / 1 mm • Height Functions: HOLD, user-defined zero setting, automatic switch-off
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.