Learn More
Seca Corp 274 Mobile stadiometer with digital display
Supplier: Seca Corp 2741900009
If you are looking for a wireless solution for taking height measurements, you cannot go wrong with the free-standing digital stadiometer seca 274. Its individual parts of high-quality aluminum can be easily assembled and tightly screwed together and its measuring range from 11 inches ‒ 7 ft 2 in can be used for small children to adults. Additional benefits include a slip resistant glass platform and anti-tip column design, white backlit display on the headpiece and wireless data transmission. • Measuring range from 11 inches ‒ 7 ft 2 in can be used for small children to adults • Slip resistant glass platform and anti-tip column design • White backlit display on the headpiece • Measuring range: 11 in – 7 ft 2 in / 30 – 220 cm • Graduation: 1/8 in /1 mm • Functions: HOLD, inch/cm switch-over, user-defined zero setting, automatic switch-off
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.