Learn More
Seca Corp 201 Retractable waist circumference measuring tape, inches-only, 0 - 80 inches
Supplier: Seca Corp 2011817009
Today’s circumference tapes also have to satisfy high medical demands. That is why seca has developed the seca 201 which can measure girth with precision. The high-quality mechanics guarantee that the 80-inch long tape unwinds easily and locks in place precisely. The housing sits comfortably and securely in the user’s hand and is tough enough to withstand accidental dropping. • Retractable measuring tape for precise body measurements in centimeters • Printed scale on side makes results easy-to-read • Compact and easy to store • Measuring range: 0 - 80 inches • Graduation: 1/8 in
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.