Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Seamless Cellulose Dialysis Tubing
Retains materials with 12,000 and higher molecular weight.
Description
- Especially useful in analytical chemistry, biology, and pharmaceutical fields
- Highly permeable to water and low molecular weight compounds
- Contains glycerin, water and 1% sulfur
- Must be stored in airtight container or rehumidified before use
- Pore diameter: 4.8nm
Specifications
Dialysis Tubing | |
45 mm | |
100 ft. | |
Glycerin, water and 1% sulfur | |
Useful in analytical chemistry, biology, and pharmaceutical fields |
Seamless Cellulose | |
1.13 in. | |
30 m | |
Each |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction