Learn More
Microsolv Technology Corporation Sealing Mat, Pre-Slit PTFE, for 96 Well Plates with 1ml Glass Inserts, 1 EA.
Supplier: Microsolv Technology Corporation 9509646
This Sealing Mat has a PTFE coating that faces the plate and seals 96 Glass Inserts in a 96 Well Plate configuration. The Mat is Pre-Slit for easy penetration by auto-injectors and the bottoms are wells for better sealing.
This item is not returnable.
View return policy