Learn More
Perkin Elmer US LLC Sealed Liquid Cell for the Variable Temperature Cell - KBr, 0.1 mm
Supplier: Perkin Elmer US LLC L1272294
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Variable Temperature Cell is used for the transmission study of solid or liquid samples at temperatures ranging from -190-250°C for solids and -70-250°C for liquids. These cells are compatible with the Frontier and Spectrum 0ne/100/400/65/Two/3 spectrometers.