Learn More
iQ Biosciences SD Rat PBMCs, 10M cells/vial
Supplier: iQ Biosciences IQBRPB102
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMCs) from the Sprague Dawley rat, a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.