Learn More
Cole-Parmer SD Mixer Controller; 120/230 VAC
Supplier:  Cole-Parmer 5500807
Encompass_Preferred
Replacement parts keep your overhead mixer running smoothlyMust be used with Mixer Motor 55008-08The precision digital controller is used to turn on and set the speed, timer, direction, and torque limits for the mixer motor.
Catalog No. NC2162488