Learn More
CHEMSCIENCE Screw Cap and PRE-SLIT Septa, Size 9-425, Red PTFE/White Silicone 1mm(0.040),CS/1000
Supplier: CHEMSCIENCE CSCS09P
Screw Cap and PRE-SLIT Septa, Size 9-425, Red PTFE/White Silicone 1mm(0.040), PK/100,CS/10PKS