Learn More
Agilent Technologies SCR CPS STEEL, MAG CAP, 100/PK
Supplier: Agilent Technologies 51882759
Screw cap, headspace, steel, magnetic cap, PTFE/silicone septa (top white, bottom blue), 18 mm, 100/pk. Cap size: 18 mm
This item is not returnable.
View return policy