Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Scott™ Escape Cylinder And Valve Assembly
The Scott™ Escape Cylinder And Valve Assembly is a compressed breathing air cylinder that is available in aluminum for infrequently-used SCBA or carbon-wrapped for daily use.
$635.00 - $1060.00
Specifications
Product Type | Air Cylinder |
---|---|
For Use With (Application) | Supplies fresh air |
For Use With (Equipment) | Self-contained Breathing Apparatus (SCBA) |
Catalog Number | Mfr. No. | Material | Duration | Pressure | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Duration | Pressure | Price | Quantity & Availability | |||
17-000-493
|
3M™
XP100321106 |
Aluminum | 5 min. |
Each for $635.00
|
|
17-000-494
|
3M™
XP100329489 |
Carbon-wrapped | 5 min. | 3000 psi |
Each for $827.50
|
|
17-000-495
|
3M™
XP100329497 |
Carbon-wrapped | 10 min. | 3000 psi |
Each for $876.00
|
|
17-000-492
|
3M™
XP100104924 |
Carbon-wrapped | 15 min. | 4500 psi |
Each for $1,060.00
|
|
Description
- 2216 psig aluminum cylinders available in 3-minute or 5-minute durations with unlimited life expectancy
- Carbon cylinders available in 3000 psig (5-minute or 10-minute durations) and 4500 psig (15-minute duration)
- 15-year life expectancy for carbon cylinders
Specifications
Air Cylinder | |
Supplies fresh air | |
Self-contained Breathing Apparatus (SCBA) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title