Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Staples Advantage Scotch™ Two-Roll Tape Dispenser

For tapes with 3 in. cores
Supplier: Staples Advantage 504936
This item is not returnable.
View return policy
Description
- Holds either one roll of tape up to 2 in. wide (51mm) or two rolls, each up to 1 in. wide (25mm)
- High-impact plastic
Specifications
2-Roll Tape Dispenser | |
1 or 2 Rolls of Tape | |
1 Roll up to 2 in. W or 2 Rolls up to 1 in. W | |
Pull and cut |
Tan | |
Plastic | |
For tapes with 3 in. cores |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title