Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Scotch-Brite™ General Purpose Scouring Pad
3M™ Medium duty scouring pad designed for everyday cleaning of cooking utensils and equipment. Scours 2 to 3 times faster and lasts 2 to 3 times longer than comparable pads. Non rusting and resilient, this synthetic pad can replace steel wool or stainless steel scrubbers.
Supplier: 3M™ 7100057473
This item is not returnable.
View return policy
Description
- Use for everyday cleaning of most cooking utensils and equipment.
- Scours 2 to 3 times faster and lasts 2 to 3 times longer than comparable pads.
- Non-rusting and resilient, this synthetic pad is an ideal replacement for steel wool or stainless steel scrubbers.
- Certified by HACCP International as 'Food Safe' and 'Fit for Purpose.'
- For fast removal of finish and proven consistent stripping
- *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved initial performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
HACCP International | |
Commercial Kitchen Cleaning | |
229 mm | |
60/Cs. | |
152 mm |
Green | |
9 in. | |
Fiber, Mineral, Resin | |
6 in. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction