Learn More
Scigen O.C.T. Compound
Water-soluble O.C.T. Compound provides specimen matrix for cryostat sectioning at -10°C and below
Supplier: Scigen 4583
This item is not returnable.
View return policy
Description
- Blue or clear compound for optimal cutting temperature
- Water-soluble blend of glycols and resins that provides convenient specimen matrix for cryostat sectioning at temperatures of -10°C and below
- Leaves no residue on slides during staining procedure, eliminating undesirable background staining
- Dissected tissue, mounted in this compound, can be frozen between -20 and -80°C
- Optimized for chromogenic IHC experiments
- Will not dull microtome knives
Specifications
Clear | |
For in vitro diagnostic use |