Learn More
Eppendorf™ Scientific Shaker Universal Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Supplier:  Eppendorf™ PTL386
 View more versions of this product
Catalog No. 14-278-074
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Requires : Clamps (sold separately)
Description
Description
  • Accept a variety of accessory clamps, test tube racks and gel trays for greater flexibility
  • Hold combinations of flasks or test tube racks
  • With pre-drilled holes
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal Platform
24 in.
610 mm
36 in.
914 mm
Innova 2350 shaker
24 x 36 in. (610 x 914 mm)
24 in.
61 cm
36 in.
91.4 cm
Pre-drilled holes
Product Suggestions
Product Suggestions