Learn More
Scientific Device Laboratory Adhesive Coated Slides
Avoid losing valuable specimens
$1279.00 - $1628.00
Catalog Number | Mfr. No. | Coating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Coating | Price | Quantity & Availability | |||||
23-769-310
|
Scientific Device
0675 |
Poly-L-Lysine |
Case of 720 for $1,628.00
|
|
|||||
23-769-311
|
Scientific Device
0685 |
Silane |
Case of 720 for $1,279.00
|
|
|||||
Description
- Adhesives will not interfere with staining
- Ideal for hybridization techniques
- Frosted ends
Specifications
1 End | |
720/Cs. |