Learn More
Scientific Device Giemsa Wright Control Slides for Malaria
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For staining smears of Plasmodium sps.
Supplier: Scientific Device 430
Description
- Control for malaria parasite staining procedure
- Scan quickly with high organism counts of red blood cells infected with Babesia sp
- Standardize and satisfy regulatory requirements for Giemsa-Wright Stain control
- Eliminate the need to maintain your own cultures
Specifications
Control Slide | |
Pk of 10 |