Learn More
Science First™ Acceleration Paradox
Shows how acceleration takes place.
Supplier: Science First™ 6111135
Includes: Two 1m boards, hinged together but separated by a dowel, a cup for the included ball, and instructions.
Description
- A ball and board are released together, the end of the board outstrips its center of mass, leaving the ball behind
- The ball then drops straight into the cup
Specifications
Acceleration Paradox | |
Shows how acceleration takes place | |
Two 1m boards, hinged together but separated by a dowel, a cup for the included ball, and instruction manual |