Learn More
Fine Science Tools Schwartz Micro Serrefines, Jaw: 10 x 1.75mm, Straight, 26 mm
Supplier: Fine Science Tools 1805201
Product: Schwartz Micro Serrefine - Straight/26mm/ Shape: Straight/ Jaw Dimensions: 10 x 1.75mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 26mm/ Serrated: No/ Unit: Each