Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ SBP Disposable Sleeve
Click to view available options
Blue
Description
- Elastic openings on both ends
- High level of comfort and breathability
- FDA approved
- One time use product
- Polypropylene disposable clothing provides affordable, lightweight protection against non-hazardous dry particulates like dirt
- Good for use in pharmaceuticals, medical. law enforcement and laboratory
Specifications
Size | Universal |
Material | Polypropylene |
Color | Blue |
Sterility | Non-sterile |
Disposable | Single-use |
Latex Free | Yes |
For Use With (Application) | Pharmaceuticals, Medical, Laboratory, Law Enforcement |
Certifications/Compliance | FDA |
Includes | 200/ case |
Length (English) | 18 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction