Learn More
Vector Laboratories Soybean Agglutinin (SBA), Unconjugated, 25mg
Supplier: Vector Laboratories L101025
SBA preferentially binds to oligosaccharide structures with terminal α- or β-linked N-acetylgalactosamine, and to a lesser extent, galactose residues. Binding can be blocked by substitutions on penultimate sugars, such as fucose attached to the penultimate galactose in blood group B substance.