Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Polycarbonate Filtration Holders

For the clarification or sterile filtration of up to about 200ml volumes of aqueous solutions

$140.70 - $199.50

Specifications

Material Polycarbonate
Material (Membrane) Hydrophilic PES
For Use With (Application) For the clarification or sterile filtration of up to about 200mL volumes of aqueous solutions
Quantity Each
Volume (Metric) Upper 250 mL
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
14-555-804
View Documents Promotion Details
Sartorius
16510
Filtration Holder
Each for $199.50
Add to Cart
 
14-555-805
View Documents Promotion Details
Sartorius
16511
Filter Holder Top Part
Each for $140.70
Add to Cart
 
Description

Description

For the clarification or sterile filtration of up to about 200ml volumes of aqueous solutions

Type 16510 is complete with receiver flask, and can be operated with sub-pressure as well as with slight over-pressure (0.5 bar is recommended for highest standing times). It is, together with a vacuum hand pump, a practical, cost-effective system for the filtration in the laboratory or outside.For sterile filtrations, the filter holder, comprised in the delivery, is equipped with a glass fibre filter 13400-0013 and enables a sterile venting for pressure compensation in order to avoid contamination of the sterile filtrate. The funnel fits onto the central opening of the lid and simplifies the pouringin of liquid in the top part. Type 16511 is like 16510, but without receiver flask. It is used on a suction flask or a vacuum manifold, e.g. Combisart.

Specifications

Specifications

Polycarbonate
For the clarification or sterile filtration of up to about 200mL volumes of aqueous solutions
250 mL
Hydrophilic PES
Each
4.7 cm
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit