Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Quantitative Grade 1288 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
$13.90 - $75.90
Specifications
Format | Disc |
---|---|
Particle Retention | 12 to 15 μm |
Filtration Speed | 10 sec. |
Grade | 1288 |
Quantity | 100/Pk. |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
14-555-078
|
Sartorius
FT3206125 |
125 mm |
Pack of 100 for $25.45
|
|
|||||
14-555-075
|
Sartorius
FT3206240 |
240 mm |
Pack of 100 for $75.90
|
|
|||||
14-555-077
|
Sartorius
FT3206150 |
150 mm |
Pack of 100 for $39.15
|
|
|||||
14-555-076
|
Sartorius
FT3206185 |
185 mm |
Pack of 100 for $48.05
|
|
|||||
14-555-080
|
Sartorius
FT3206090 |
90 mm |
Pack of 100 for $16.07
|
|
|||||
14-555-079
|
Sartorius
FT3206110 |
110 mm |
Pack of 100 for $18.67
|
|
|||||
14-555-081
|
Sartorius
FT3206070 |
70 mm |
Pack of 100 for $14.78
|
|
|||||
14-555-082
|
Sartorius
FT3206055 |
55 mm |
Pack of 100 for $13.90
|
|
|||||
14-555-083
|
Sartorius
FT3206047 |
47 mm |
Pack of 100 for $14.78
|
|
|||||
14-555-073
|
Sartorius
FT3206500 |
500 mm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
14-555-074
|
Sartorius
FT3206400 |
400 mm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.Specifications
Disc | |
10 sec. | |
100/Pk. |
12 to 15 μm | |
1288 | |
0.21 mm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title