Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Microsart™ e.motion Membrane Filters
For colony counting, particle testing and microscopy. Specifically designed for use in the Microsart™ e.motion dispenser, the filters are individually sterile-sealed in a format packaged on a pleated band, without protective paper. Can also be used with Sartorius nutrient pad sets.
Supplier: Sartorius 114H6Z47SFM
Description
Microsart™ e.motion membrane filters are suitable for the use with the Microsart™ e.motion dispenser. The filters are individually sterile-sealed on a band without any protective paper. The special pleating as well as the shape of the sealing of the band of membrane filters ensures a perfectly flat and uniform dispensing of the individual membrane filters. The Microsart™ e.motion membrane filters are also ideal for all users of the Sartorius Nutrient Pad Sets: most of the NPS types are available with Microsart™ e.motion membrane filters. Outstanding recovery rates for microorganisms 0.45 μm are acc. to ISO 7704. Multi-fit: Fits into other dispensers Protective paper-free Packaged on a special pleated band Product data is printed on individual envelopes. High flow membranes. Gamma irradiated, 25 kGray Design 47 or 50 mm in diameter, white, grey or green and gridded Growth Promotion Test acc. to ISO 7704 no enhancement or inhibition by the grid lines no enhancement or inhibition due to chemical extractables no enhancement or inhibition by the sterilization process.Specifications
White | |
0.45 μm | |
For colony count; sterility testing; particle count; microscopy | |
450/Pk. |
Cellulose Nitrate | |
Sterile | |
Black | |
47 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction