Learn More
Sartorius Arium™ Bagtank Rollers
Arium™ Bagtank Rollers enable Arium™ Bagtanks to be moved quickly and conveniently to wherever it is needed. Arium™ Bagtank Rollers are an optional accessory designed for use with Arium™ Bagtank 50 and Arium™ Bagtank 100 purified water storage units.
Supplier: Sartorius H2OATR
Description
The closed Arium™ Bagtank system reliably protects purified water from secondary contamination to ensure consistently high water quality. Unlike a standard tank system, the bag can be replaced in less than 5 minutes and eliminates time-consuming cleaning procedures with hazardous chemicals.
- Optional rollers makes bagtanks extremely portable and flexible
- Used with the Arium™ Bagtank 50 or Bagtank 100
- Compatible with Arium™ Comfort and Arium™ Advance Water Purification Systems
Specifications
Arium Bagtank 50 and Bagtank 100 Ultrapure Water Storage System | |
Rollers | |
Storage of ultra-purified water | |
4 Rollers and Fittings |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.