Learn More
Sartorius arium™ ion exchange Resin
Supplier: Sartorius 615DIC01
Description
arium™ refill pack of ion exchange resin mix in 25l PE bag for complete deionization cartridges
Specifications
Ion Exchange Resin | |
Remove cations and anions from the water by exchanging H+ and OH- |