Learn More
Sartorius Actiosart Syringe Filters
For ultra-cleaning of gases e.g. to absorb unpleasant odors
Supplier: Sartorius 17840Q
Description
- MBS housing with high-adsorptive active carbon filling, connector on both sides male Luer slip
- Sample cleaning and particle removal
- Easy to handle
- 100% tested during manufacture
Specifications
Single-use | |
PTFE | |
Syringe Filter |