Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Qualitative Grade 34/N Filter Papers

This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.

$185.00 - $238.00

Specifications

Format Folded
Grade 34/N
Quantity 100/Pk.
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
14-555-003
View Documents Promotion Details
Sartorius
FT4478125
125 mm
Pack of 100 for $185.00
Add to Cart
 
14-555-002
View Documents Promotion Details
Sartorius
FT4478320
320 mm
Pack of 100 for $238.00
Add to Cart
 
Description

Description

Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.
Specifications

Specifications

Folded
34/N
100/Pk.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit