Learn More
LumiraDx SARS-CoV-2 Surveillance Test
LumiraDx SARS-CoV-2 Ag Surveillance Test Kit (48/Pack) is used in conjunction with the LumiraDx Instrument allowing for users to perform COVID-19 surveillance tests. Up to 5 nasal swab specimens can be pooled for testing, aiding in the surveillance of COVID-19 in populations.
Supplier: LumiraDx L016000312048
Description
- The LumiraDx SARS-CoV-2 Antigen (Ag) Surveillance Tests are used with the LumiraDx Instrument for COVID-19 surveillance testing.
- With lab-comparable results in 12 minutes, the LumiraDx SARS-CoV-2 Ag Surveillance Test delivers fast, accurate, cost-efficient pooled testing onsite to verify that COVID-19 isn’t spreading in your population or quickly implement preventative measures when clusters emerge.
- For Surveillance use only*
- 100.0% positive agreement and 96.6% negative agreement with LumiraDx SARS-CoV-2 Ag Test
- Up to 5 specimens on a single Test Strip
- Pooled swab sample may be applied to a single test strip, without sacrificing sensitivity
- Results in 12 minutes
- Orders require executed contractual agreement. Please contact your Fisher Healthcare Account Representative for more information
- *Surveillance Testing is intended to monitor community - or population - level outbreaks of disease, or to characterize the incidence and prevalence of disease. It is performed on de-identified specimens, and thus, results are not linked to individual people. The LumiraDx SARS-CoV-2 Ag Surveillance Test has not been cleared, approved, or authorized by FDA. This test should be used for surveillance purposes (i.e., to inform population or community-level decision-making on de-identified specimens) only. FDA does not regulate surveillance tests.
Specifications
Yes | |
SARS-CoV-2 | |
No | |
Nasal or Nasopharyngeal Swab Specimens | |
Surveillance Use Only | |
12 min. |
Not Applicable | |
Microfluidic Immunofluorescence | |
Test Strips | |
96.6% to LumiraDx Ag Test | |
LumiraDx Instrument |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Surveillance Testing is intended to monitor community - or population - level outbreaks of disease, or to characterize the incidence and prevalence of disease. It is performed on de-identified specimens, and thus, results are not linked to individual people. The LumiraDx SARS-CoV-2 Ag Surveillance Test has not been cleared, approved, or authorized by FDA. This test should be used for surveillance purposes (i.e., to inform population or community-level decision-making on de-identified specimens) only. FDA does not regulate surveillance tests.