Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ SARS-CoV-2 Process Control
SARS-CoV-2 Process Control can be used for SARS-CoV-2 assay development, full process control (including extraction), sample adequacy control and spike-in controls.
Specifications
Level | Biosafety Level 1 |
---|---|
Packaging | Plastic Box; Individually packaged contents |
Quantity | 6/Pk. |
Source | United States |
Form | Lyophilized |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
23-047-375
|
Microbiologics™
R9000 |
Lyophilized Pellets |
Pack of 6 for $787.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-047-376
|
Microbiologics™
R9001 |
Lyophilized Swabs |
Pack of 6 for $762.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- SARS-CoV-2 Process Control is made with RNA transcripts containing diagnostically relevant targets (CDC and WHO consensus sequences) encapsulated in a phage protein.
- Nucleotides contained in each pellet/swab (All positions in reference to GenBank Acc. NC_045512.2)
- Orf1ab/RdRP: 12,690 ..12,797 | 13,342 ..13,460 | 14,080 ..14,186 | 15,431 ..15,530
- S (Spike): 18,778 ..18,909 | 24,354 ..24,900
- E (Envelope): 26,245 ..26,427 | 26,269 ..26,381
- N (Nucleocapsid): 28,237 ..29,280
- Includes human respiratory epithelial cells
- RUO
Specifications
Biosafety Level 1 | |
6/Pk. | |
Lyophilized |
Plastic Box; Individually packaged contents | |
United States |
Safety and Handling
ShelfLife : 8 Months (Minimum)
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title