Learn More
Zeptometrix SARS-CoV-2 Lineage B.1.351; Beta Variant (South_Africa/KRISP-K005325/2020) Culture Fluid (UV Inactivated)
Infectious virus preparation designed to meet research and product development needs.
Supplier: Zeptometrix 0810613UV
Description
SARS-Related Coronavirus 2 Lineage B.1.351; Beta Variant (Isolate: South_Africa/KRISP-K005325/2020) is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus. Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2), isolate hCoV-19/South Africa/KRISPK005325/2020 (also referred to as 501Y.V2.HV and 501Y.V2.HV001) was isolated from an oropharyngeal swab from a 40-year-old human male in Ugu district, KwaZulu-Natal, South Africa on November 16, 2020. Under the nomenclature system introduced by GISAID (Global Initiative on Sharing All Influenza Data), SARSCoV-2, isolate hCoV-19/South Africa/KRISPK005325/2020 is assigned lineage B.1.351 and GISAID clade GH using Phylogenetic Assignment of Named Global Outbreak LINeages (PANGOLIN) tool. The complete genome of the clinical isolate of SARS-CoV-2, hCoV-19/South Africa/KRISP-K005325/2020 has been sequenced (GISAID: EPI_ISL_678615). Each frozen aliquot contains 1 mL of UV inactivated, clarified viral culture fluid. The pre-inactivation titer was determined from an infectious aliquot. Viral inactivation is verified after UV inactivation by the absence of viral growth in tissue culture-based infectivity assays.Specifications
ISO 13485 | |
Culture Fluid | |
1 mL |
Store at -65°C or below | |
Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For the purchase of Zeptometrix's BSL2 or BSL3 level products the Laboratories Biosafety Level must be consistent with the products requirement.