Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix SARS-CoV-2 (Isolate: USA-WA1/2020) Lysate
SARS-CoV-2 (Isolate: USA-WA1/2020) Culture Fluid is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus.
Specifications
Content And Storage | Recommended storage temperature -65°C or below |
---|---|
Type | Lysate |
Intended Use | For research or further manufacturing only. Not for use in diagnostic procedures |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-158-376
|
ZeptoMetrix
08105870.5MG |
0.5 mg |
Each for $963.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-158-375
|
ZeptoMetrix
0810587 |
1 mg |
Each for $1,899.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- This product is intended for research, product development, quality assurance testing, or further manufacturing use.
- Viral lysates can be used as an antigen, as a source for the purification of viral proteins, or in assays for the detection of viral antibodies.
Specifications
Recommended storage temperature -65°C or below | |
Lysate | |
For research or further manufacturing only. Not for use in diagnostic procedures |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title