Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zeptometrix SARS-CoV-2 (Isolate: Italy-INMI1) Culture Fluid (UV Inactivated)
Infectious virus preparation designed to meet research and product development needs.
Supplier: Zeptometrix 0810589UV
Description
UV inactivated viral culture fluids are sold as consumable testing materials. The suitability and performance characteristics should be determined by your laboratory for each intended usage. These products are NOT intended for use in the manufacture or processing of injectable products subject to licensure under section 351 of the Public Health Service Act or for any other product intended for administration to humans.Specifications
ISO 13485 | |
Culture Fluid | |
1 mL |
Store at -65°C or below | |
Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For the purchase of Zeptometrix's BSL2 or BSL3 level products the Laboratories Biosafety Level must be consistent with the products requirement.
Spot an opportunity for improvement?
Provide Content Correction