Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sarkosyl, Fisher BioReagents
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Description
- Sarkosyl is an anionic detergent used for cell lysis during the extraction of RNA
- Also suitable for the solubilization of membrane protein
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Physical Form | Solid |
Melting Point | 45°C |
Assay Percent Range | ≥95 % |
Linear Formula | NaO2CCH2N(CH3)CO(CH2)10CH3 |
pH | 7.5 to 8.5 |
Quantity | 500 g |
Merck Index | 15, 4401 |
Identification | Pass Test |
Formula Weight | 293.38 |
Show More |
Safety and Handling
![](http://beta-static.fishersci.com/images/Gray.gif)
Hazard Statement | CAUTION! |
Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
Recommended Storage | RT |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction