Learn More
Electron Microscopy Sciences Optical Fiber Sapphire Cleaver Tool
Supplier: Electron Microscopy Sciences 62165SC
Retractable, ground and polished natural sapphire wedge with a 60 degree included angle. Retained in a quality metal pen like casing with pocket clasp. Sapphire is 2.0mm wide x 3.0mm long, overall length is 133mm.